Learn More
Air Impurities Removal Systems Extract-ALL™ Portable Fume Extraction System: Activated Carbon Refill
Carbon refill for F-987-2A and F-987-4A
Manufacturer:  Air Impurities Removal Systems RF9874C
Catalog No. 19-097-447
Add to cart
Description
Description
  • For use with Extract-ALL Portable Fume Extraction Systems
Specifications
Specifications
Carbon
Safety
F-987-2A and F-987-4A filters for compact air cleaning system
1.81kg
Captures volatile solvents, touch-up paint fumes and other noxious odors
Filter
5 lb.