Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Extran™ MA 05

Use for manual cleaning. MilliporeSigma Extran™ MA 05 is an alkaline concentrated liquid that is phosphate free. It is not for alkali-sensitive material (e.g. aluminum). Extran products are biodegradable, with no residues, impurities or harsh chemica

$57.43 - $315.41

Specifications

pH Range 13 (in water, 20°C)
Format Concentrated liquid
For Use With (Application) Manual cleaning
Color Colorless to light yellow
Container Type Plastic
View More Specs

Products 4
Catalog Number Mfr. No. Size Price Quantity & Availability  
Catalog Number Mfr. No. Size Price Quantity & Availability  
M1400002500 View Documents MilliporeSigma™
1.40000.2500
2.5L Each for $57.43
Add to cart
 
M1400005000 View Documents MilliporeSigma™
1.40000.5000
5L Each for $91.50
Add to cart
 
M1400009010 View Documents MilliporeSigma™
1.40000.9010
10L Each for $163.41
Add to cart
 
M1400009025 View Documents MilliporeSigma™
1.40000.9025
25L Each for $315.41
Add to cart
 
Specifications

Specifications

13 (in water, 20°C)
Manual cleaning
Plastic
Concentrated liquid
Colorless to light yellow
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit