Learn More
Fisherbrand™ Extreme Thermal Box Liners
Fisherbrand Extreme Thermal Box Liners feature a triple-layer of thermal protection that will keep sensitive products at the right temperature during transit.
 View more versions of this product
Catalog No. 07-100-102
Add to cart
Description
Description
Features:
  • Cold chain protection for laboratory products and pharmaceutical during transit
  • Thick 1-inch thermal foam cushions insulating material
  • FDA compliant material
  • Use with cold packs or dry ice for extended transit times
  • Liners are vented for using dry ice
  • Box not included fits standard cube size box
  • Made in USA
  • Space saving one piece design, stores flat saving storage space over traditional EPS coolers
  • Superior insulation properties and thinner walls allow for more payload space
Order Info
Case quantity only 20 per case
Warranty and Services
Warranty against defects in material or workmanship.
Specifications
Specifications
White Tape
23in x 23in
584mm
20
12 Cube
Metalized PET Silver Exterior
51mm
Outer Layer: Metalized PET laminated to PE, Middle Layer: Open Cell PU Foam, Inner Layer: White LDPE (FDA approved for incidental food contact).
548mm