Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ EYA2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA555487
Description
Immunogen sequence: GTTGFYQGGN GLGNAAGFGS VHQDYPSYPG FPQSQYPQYY GSSYNPPYVP ASSICPSPLS TSTYVLQEAS HNVPNQSSES LAGEYNTHNG PSTPAKEGDT DRPHRASDGK LRGRSKRSSD PSPAGDNEIE RVFVWDLD Highest antigen sequence identity to the following orthologs: Mouse - 86%, Rat - 88%.
Functions both as protein phosphatase and as transcriptional coactivator for SIX1, and probably also for SIX2, SIX4 and SIX5. Tyrosine phosphatase that dephosphorylates 'Tyr- 142' of histone H2AX (H2AXY142ph) and promotes efficient DNA repair via the recruitment of DNA repair complexes containing MDC1. 'Tyr-142' phosphorylation of histone H2AX plays a central role in DNA repair and acts as a mark that distinguishes between apoptotic and repair responses to genotoxic stress. Its function as histone phosphatase may contribute to its function in transcription regulation during organogenesis. Plays an important role in hypaxial muscle development together with SIX1 and DACH2; in this it is functionally redundant with EYA1.
Specifications
EYA2 | |
Polyclonal | |
Unconjugated | |
EYA2 | |
Drosophila-type eyes absent 2-like protein; Eab1; EYA transcriptional coactivator and phosphatase 2; Eya2; eyes absent 2; eyes absent 2 homolog; eyes absent 2 homolog (Drosophila); eyes absent 2 isoform a; eyes absent homolog 2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
2139 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
O00167 | |
EYA2 | |
Recombinant protein corresponding to Human EYA2. Recombinant protein control fragment (Product #RP-91554). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction