Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EYA3 Rabbit anti-Human, Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$138.37 - $320.71

Specifications

Antigen EYA3
Applications Immunofluorescence, Western Blot
Conjugate Unconjugated
Format Purified
Host Species Rabbit
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB093777
View Documents
Novus Biologicals
NBP304637100UL
100 μg
Each for $320.71
Add to cart
 
NB093778
View Documents
Novus Biologicals
NBP30463720UL
20 μg
Each for $138.37
Add to cart
 
Description

Description

EYA3 Polyclonal antibody specifically detects EYA3 in Human, Rat samples. It is validated for Western Blot, Immunofluorescence
Specifications

Specifications

EYA3
Unconjugated
Rabbit
Vision
PBS with 50% glycerol, pH7.3.
2140
IgG
Primary
Store at -20°C. Avoid freeze-thaw cycles.
Immunofluorescence, Western Blot
Purified
RUO
Human, Rat
DKFZp686C132, EC 3.1.3.48, eyes absent (Drosophila) homolog 3, eyes absent 3, eyes absent homolog 3, eyes absent homolog 3 (Drosophila)
Recombinant fusion protein containing a sequence corresponding to amino acids 1-300 of human EYA3 (NP_001981.2).
Polyclonal
Affinity purified
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit