Learn More
Fisherbrand Eyeshield
Optical-grade lens curves around the face to maximize protection to the front and sides of the eyes
Description
Optical-grade lens curves around the face to maximize protection to the front and sides of the eyes
- For use with Fisherbrand Resposables EyeShields Frames (cat. no. 23500926)
- Allow easy distribution where needed of disposable EyeShields replacement lenses
- Ten envelopes of 25 disposable, clear lenses
Compliance
Meet OSHA requirements and EEC mandates
Specifications
Provides excellent visibility while protecting the face and neck from accidental splashes | |
Yes | |
Optical-grade |
50/Pk. | |
Clear | |
Eyeshield |