Learn More
Sklar InstrumentsSupplier Diversity Partner Forceps, Eye Tissue Forceps, Sklar 4 in., 1x2 Teeth, Straight, Stainless Steel, EA. DFS Item
Manufacturer:  Sklar InstrumentsSupplier Diversity Partner 663240
Forceps, Eye Tissue Forceps, Sklar, 4 in., 1x2 Teeth, Straight, Stainless Steel, EA. Sklar Iris Tissue Forceps are most commonly used for grasping and handling tissue in ophthalmic procedures. The narrow tips, however, allow this instrument to be beneficial for use in other microsurgical procedures as well. Iris Tissue Forceps are available with 1X2 teeth in multiple lengths and curvatures. This product is straight with 1 X 2 teeth and 4 inches in length. Lifetime warranty against all manufacturer defects.
Catalog No. 50-111-8917