Learn More
Thermo Scientific™ EZ-Link™ Sulfo-NHS-LC-Biotin and Labeling Kits
Biotinylate protein amines (e.g., lysines) with this very popular mid-length (22.4A), water-soluble, NHS-ester biotinylation reagent for antibody or cell-surface labeling.
Specifications
Cleavable No
Membrane Permeable No
Molecular Weight 556.59
PEGylated No
Spacer Arm Length 22.4
View More Specs
Products 1
Catalog Number Mfr. No. Description Price Quantity & Availability  
Catalog Number Mfr. No. Description Price Quantity & Availability  
PI21327
View Documents
Thermo Scientific™
21327
EZ-Link Sulfo-NHS-LC-Biotin, No-Weigh Format This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description
Description
Thermo Scientific™ EZ-Link Sulfo-NHS-LC-Biotin is an intermediate-length, water-soluble biotinylation reagent for labeling antibodies, proteins and other molecules that have primary amines.
Highlights:
  • Water-soluble analog of NHS-LC-Biotin
  • Reacts with primary amines at pH 7-9 in non-amine-containing buffers such as borate, carbonate or phosphate buffers
  • Negative charge of -SO3 group keeps biotinylation localized at the cell surface
  • Creates an amide bond
U.S. Patents 5,892,628 and 5,872,261.
Specifications
Specifications
No
556.59
22.4
Yes
No
No
-20°C with desiccant