Learn More
Thermo Scientific™ EZ-Link™ Sulfo-NHS-SS-Biotin, No Weigh Format
Thermo Scientific™ EZ-Link™ Sulfo-NHS-SS-Biotin No-Weigh™ Format is a water-soluble, NHS-ester biotinylation reagent whose spacer arm includes a cleavable disulfide bond for reversible labeling of proteins and cell surface primary amines.
Manufacturer:  Thermo Scientific™ 21328
 View more versions of this product
Catalog No. PI21328
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description
Description
Thermo Scientific™ EZ-Link Sulfo-NHS-SS-Biotin enables simple and efficient biotin labeling of antibodies, proteins and any other primary amine-containing biomolecules in solution.
Highlights:
  • Cleavable analog of Sulfo-NHS-LC-Biotin
  • Can remove biotin label by cleaving spacer disulfide bond with reducing agents, such as DTT
  • Ideal for affinity purification experiments that require efficient elution from immobilized avidin supports (cleaving disulfide bond releases proteins)
  • Long spacer arm reduces steric hindrance for binding interactions
Specifications
Specifications
Amine
EZ-Link Sulfo-NHS-SS-Biotin, No-Weigh Format
606.69
No
8 x 1mg
-20°C protected from moisture
Yes
Sulfo-NHS ester of biotin with an ethyl-1,3-dithioproprionate spacer
No
24.3
8 uses to label a total of 88mg of IgG using typical conditions
Yes