Brady™ EZ Panel Loc Breaker Lockouts
Manufacturer: Brady™ 65815
 View more versions of this product
Catalog No. 19-103-958
  Check Availability
Add to Cart
    Specifications
    Description The EZ Panel Loc System includes lockout devices, accommodating an exceptionally wide range of breaker models. The Snap-on breaker is for use on Square D NQO and other breakers that have a hole in the switch tongue. Just align pins with hole and snap closed. Pins prevent switch from being thrown.
    Dimensions (L x W x H) 0.90 in. x 1.88 in. x 0.74 in.
    Quantity 6/Package
    Color Red
    Product Type Snap-on
    • The EZ Panel Loc System includes lockout devices, accommodating an exceptionally wide range of beaker models
    • Snap-on and clamp-on breaker lockout options accommodate almost any situation
  • Description & Specifications
Related Products
Product Content Feedback