Learn More
Innovating Science™ EZ Prep Chemicals (A-M) Products for Science Education
Capsules are pre-measured to make a chemical solution.
Manufacturer:  Aldon Corporation IS4053
 View more versions of this product
Catalog No. S4053A
Add to cart
Description
Description
  • Add to distilled water and stir to make the amount of solution indicated
  • There is no need to handle chemicals as the chemicals are contained within the capsule
  • Decrease preparation time since no weighing of chemicals is required
  • Decrease disposal costs because no extra chemicals remain
Specifications
Specifications
Basic Fuschin 1%
English
Preparations of chemical solutions
Chemistry
White
7
Lab
Capsule
1 Capsule