Learn More
Innovating Science™ EZ Prep Chemicals (A-M) Products for Science Education
Capsules are pre-measured to make a chemical solution.
Manufacturer:  Aldon Corporation IS4018
 View more versions of this product
Catalog No. S96675
Add to cart
This item is not returnable. View return policy
Description
Description
  • Add to distilled water and stir to make the amount of solution indicated
  • There is no need to handle chemicals as the chemicals are contained within the capsule
  • Decrease preparation time since no weighing of chemicals is required
  • Decrease disposal costs because no extra chemicals remain
Specifications
Specifications
14
English
Capsule
Preparations of chemical solutions
White
7
Methyl Orange 0.01%
Chemistry
1 Capsule