Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Innovating Science™ EZ Prep Chemicals (A-M) Products for Science Education

Capsules are pre-measured to make a chemical solution.

$5.15 - $16.25

Specifications

Category Chemistry
Format Powder
For Use With (Application) Preparations of chemical solutions
View More Specs

Products 19
Catalog Number Mfr. No. Product Type Price Quantity & Availability  
Catalog Number Mfr. No. Product Type Price Quantity & Availability  
S96664 Products for Science Education
View Documents
Aldon Corporation
IS4007
Benedicts Reagent
Each for $9.80
Add to cart
 
S96675 Products for Science Education
View Documents
Aldon Corporation
IS4018
Methyl Orange 0.01%
Each for $16.25
Add to cart
 
S4049A Products for Science Education
View Documents
Aldon Corporation
IS4049
Methylene Blue 1%
Each for $6.40
Add to cart
 
S4050A Products for Science Education
View Documents
Aldon Corporation
IS4050
Methylene Blue 0.1%
Each for $5.20
Add to cart
 
S4051A Products for Science Education
View Documents
Aldon Corporation
IS4051
Methyl Green 1%
Each for $7.25
Add to cart
 
S4052A Products for Science Education
View Documents
Aldon Corporation
IS4052
Malachite Green 1%
Each for $5.15
Add to cart
 
S4054A Products for Science Education
View Documents
Aldon Corporation
IS4054
Eosin Y 1%
Each for $5.15
Add to cart
 
S4053A Products for Science Education
View Documents
Aldon Corporation
IS4053
Basic Fuschin 1%
Each for $5.15
Add to cart
 
S4055A Products for Science Education
View Documents
Aldon Corporation
IS4055
Crystal Violet 0.1%
Each for $5.15
Add to cart
 
S4056A Products for Science Education
View Documents
Aldon Corporation
IS4056
Crystal Violet 1%
Each for $5.15
Add to cart
 
S4060A Products for Science Education
View Documents
Aldon Corporation
IS4060
Methyl Orange 0.1%
Each for $5.20
Add to cart
 
S4059A Products for Science Education
View Documents
Aldon Corporation
IS4059
Bromothymol Blue 0.5%
Each for $5.20
Add to cart
 
S4057A Products for Science Education
View Documents
Aldon Corporation
IS4057
Bromothymol Blue 0.1%
Each for $5.70
Add to cart
 
S4062A Products for Science Education
View Documents
Aldon Corporation
IS4062
Bromocresol Green 0.4%
Each for $5.20
Add to cart
 
S4058A Products for Science Education
View Documents
Aldon Corporation
IIS4058
Bromothymol Blue 0.04%
Each for $5.20
Add to cart
 
S4063A Products for Science Education
View Documents
Aldon Corporation
IS4063
Bromocresol Purple 0.4%
Each for $5.70
Add to cart
 
S4064A Products for Science Education
View Documents
Aldon Corporation
IS4064
Congo Red 0.1%
Each for $5.15
Add to cart
 
S4065A Products for Science Education
View Documents
Aldon Corporation
IS4065
Cresol Red 0.2%
Each for $5.20
Add to cart
 
S05890 Products for Science Education
View Documents
Aldon Corporation
ADHH0071CONC
Hydrochloric Acid 1M
Each for $5.70
N/A
Due to product restrictions, please to purchase or view availability for this product.
Description

Description

  • Add to distilled water and stir to make the amount of solution indicated
  • There is no need to handle chemicals as the chemicals are contained within the capsule
  • Decrease preparation time since no weighing of chemicals is required
  • Decrease disposal costs because no extra chemicals remain

Specifications

Specifications

Chemistry
Powder
Preparations of chemical solutions
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit