Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Microbiologics™ Geobacillus stearothermophilus derived from ATCC 12980

EZ-Accu Shot provides a convenient, reliable solution for Growth Promotion Testing, Media Challenge Testing and more. Each instant-dissolve microorganism pellet is designed to deliver 10-100 CFU per inoculum. Provides 8 hours of stability after rehydration. Meet Pharmacopeia guidelines with ease.

Manufacturer:  Microbiologics™ 0872A

Catalog No. 10-810-313


Due to product restrictions, please to purchase or view availability for this product.

This item is not returnable. View return policy

Description

Description

  • Biosafety Level: 1
  • Quantitative microorganism preparation that delivers less than 100 CFU per 0.1 ml of hydrated suspension.
  • No dilutions required.
  • Kit contains: 5 pellets of a single enumerated strain and 5 vials of hydrating fluid (1.2 ml per vial).
Specifications

Specifications

1
5 pellets
Plastic Box
ATCC 12980
2 to 8°C
EZ-Accu Shot
United States
8 Months (Minimum)
EZ-Accu Shot
Growth Promotion Testing
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit