Learn More
BioVision IncSupplier Diversity Partner EZCellTM Cell Cycle Analysis Kit: Highly Sensitive, HTS compatible, Flow-cytometry assay to identify cells at different stages of the Cell Cycle. 100 Assays DFS Item
Manufacturer:  BioVision IncSupplier Diversity Partner K920100
EZCellTM Cell Cycle Analysis Kit: Cell cycle is a ubiquitous, complex sequence of events leading to growth and proliferation of cells. Anomalies in cell cycle progression can lead to tissue hyperplasia and diseases such as cancer. Cell cycle can be subdivided into interphase (G0/G1, S and G2) and mitotic (M) phase (prophase, metaphase, anaphase and telophase). BioVision's Cell Cycle Analysis Kit provides a quick and easy method to detect the number of cells in a cell population, which are at a specific stage of the cell cycle. Our kit utilizes a nuclear dye, the binding of which to nucleic acids in the cell results in fluorescence signal, which is proportional to cellular DNA content. The percentages of cells in different phases of the cell cycle (G0/G1, S, and G2/M) can be quantified by flow cytometry. Our method is non-radioactive, rapid and accurate and can be used for high throughput cell cycle analysis with contemporary flow cytometer instruments.
Catalog No. 50-111-9987