Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Moore Medical EZe-Band™ Latex-Free Elastic Bandage

Reusable, self-closing wrap provides orthopedic-grade compression and support

Manufacturer:  Moore Medical 63369

 View more versions of this product

Catalog No. 19-898-300

This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

  • Unique knitted construction provides superior absorbency and breathability
  • Double closure delivers maximum control and consistent compression
  • No pins, clips or tape required
  • Sterility assured by Tyvek™ pouch

For use in Hospitals, Physician Offices, Industrial Health, Sports Medicine, Long Term Care, Home Health, School Health and Burn Units. Helps prevent possible future latex sensitization.

Specifications

Specifications

Non-sterile
4.6m
Double self
3 in.
Yes
Elastic
5 yd.
Self adhesive
EZe-Band™ Bandage
7.6cm
For non-slip compression
5 yd. x 3 in. (4.6m x 7.6cm)
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit