Learn More
Abnova™ EZH2 293T Cell Transient Overexpression Lysate (Denatured) (T03)
Human overexpression lysate
Manufacturer:  Abnova™ H00002146T03
Catalog No. 89-943-786
Add to cart
Specifications
Specifications
Western Blot
293T
NM_004456
pCMV-EZH2 full-length
100 uL
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Human
EZH2 293T Cell Transient Overexpression Lysate(Denatured)
Theoretical M.W. 82.72kDa
Transient overexpression cell lysate was tested with anti-EZH2 antibody (Mfr. No. H00002146-D02P) by Western Blot.
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)