Learn More
EZH2 Goat anti-Human, Mouse, Polyclonal, R&D Systems™
Goat Polyclonal Antibody
Manufacturer:  R&D Systems AF4767SP
Catalog No. AF4767SP
Add to cart
This item is not returnable. View return policy
Description
Description
EZH2 Polyclonal antibody AF4767SP specifically detects EZH2 in Human, Mouse samples. It is validated for Immunocytochemistry, Western Blotting
Specifications
Specifications
EZH2/KMT6
LYOPH
Western Blot 1 ug/mL, Immunocytochemistry 5-15 ug/mL
Q15910
EC 2.1.1, EC 2.1.1.43, enhancer of zeste (Drosophila) homolog 2, enhancer of zeste 2, Enhancer of zeste homolog 2, enhancer of zeste homolog 2 (Drosophila), ENX1, ENX-1KMT6EZH1KMT6A, histone-lysine N-methyltransferase EZH2, KMT6, Lysine N-methyltransferase 6, MGC9169
Goat
IgG
25 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects human and mouse EZH2 in direct ELISAs and Western blots.
Human, Mouse
Western Blot, Immunocytochemistry
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
EZH2
E. coli-derived recombinant human EZH2 Gly512-Ile645 Accession # Q15910
Antigen Affinity-purified
RUO
Primary
2146
Reconstitute at 0.2 mg/mL in sterile PBS.