Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EZH2/KMT6 Mouse anti-Human, Clone: EZH2/2536, Novus Biologicals™

Mouse Monoclonal Antibody

$138.37 - $352.89

Specifications

Antigen EZH2/KMT6
Clone EZH2/2536
Concentration 0.2 mg/ml
Dilution Western Blot 1-2 ug/ml, Protein Array
Applications Western Blot, Peptide Array
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB121847
View Documents
Novus Biologicals
NBP30718710UG
100 μg
Each for $352.89
Add to cart
 
NB121848
View Documents
Novus Biologicals
NBP30718720UG
20 μg
Each for $138.37
Add to cart
 
Description

Description

EZH2/KMT6 Monoclonal antibody specifically detects EZH2/KMT6 in Human samples. It is validated for Western Blot, Protein Array
Specifications

Specifications

EZH2/KMT6
0.2 mg/ml
Western Blot, Peptide Array
Purified
RUO
Human
EC 2.1.1, EC 2.1.1.43, enhancer of zeste (Drosophila) homolog 2, enhancer of zeste 2, Enhancer of zeste homolog 2, enhancer of zeste homolog 2 (Drosophila), ENX1, ENX-1KMT6EZH1KMT6A, histone-lysine N-methyltransferase EZH2, Lysine N-methyltransferase 6, MGC9169
Recombinant full-length human EZH2/KMT6 protein
Monoclonal
Protein A or G purified
EZH2/2536
Western Blot 1-2 ug/ml, Protein Array
Unconjugated
Mouse
Breast Cancer, Cancer, Chromatin Research, Phospho Specific, Prostate Cancer
10 mM PBS with 0.05% BSA
2146
IgG2a Kappa
Primary
Store at 4°C.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit