Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EZH2 Mouse anti-Human, Mouse, Rat, Clone: AC22, eBioscience™ Non-distribution product as customer accommodation.

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen 14986782

 View more versions of this product

Catalog No. 50-112-2893


Add to cart

Description

Description

Description: The monoclonal antibody AC22 recognizes EZH2 (Enhancer of zeste homolog 2), a member of the polycomb group (PcG) proteins responsible for repression of gene expression through recognition and modification of histone methylation and chromatin structure. EZH2 comprises the catalytic subunit of the polycomb repressive complex 2 (PRC2) and directly represses gene expression through methylation of histone H3 on lysine 27 (H3K27) as well as direct protein targets such as GATA4. The PRC2 complex gene repression is necessary for proper development and differentiation. Underexpression of EZH2 is linked to abnormal developmental patterning and a loss of stem cell pluripotency, while overexpression of EZH2 is present in many tumors and correlates with poor prognosis in melanoma, breast, and prostate cancer. Applications Reported: This AC22 antibody has been reported for use in immunoprecipitation, western blotting, and immunocytochemistry. Applications Tested: This AC22 antibody has been tested by immunocytochemistry of methanol-fixed cells and can be used at less than or equal to 5 µg/mL. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Purity: Greater than 90%, as determined by SDS-PAGE. Aggregation: Less than 10%, as determined by HPLC. Filtration: 0.2 µm post-manufacturing filtered.

EZH2 (Histone-lysine N-methyltransferase) is a member of the Polycomb-group (PcG) family. PcG family members form multimeric protein complexes, which are involved in maintaining the transcriptional repressive state of genes over successive cell generations. This protein associates with the embryonic ectoderm development protein, the VAV1 oncoprotein, and the X-linked nuclear protein. EZH2 may play a role in the hematopoietic and central nervous systems. Mutations affecting EZH2 can cause Weaver syndrome (WVS).
Specifications

Specifications

EZH2
Monoclonal
0.5 mg/mL
PBS with 0.09% sodium azide; pH 7.2
Q61188, Q15910
Ezh2
IgG2a, kappa
100 μg
4° C
Primary
14056, 2146, 312299
Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blot
AC22
Unconjugated
EZH2
Liquid
Mouse
Affinity chromatography
RUO
Antibody
Monoclonal
Human, Mouse, Rat
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only.