Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Lab Vision™ Ezrin/p81/80K/Cytovillin Ab-1, Mouse Monoclonal Antibody

Specifically detect ezrin in human, monkey, cow, kangaroo, rat, mouse and hamster samples with Thermo Scientific™ Ezrin/p81/80K/Cytovillin Ab-1, Mouse Monoclonal Antibody.

Specifications

Antigen Ezrin/p81/80K/Cytovillin Ab-1
Clone 3C12
Applications Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Monoclonal
Host Species Murine
View More Specs

Products 4
Catalog Number Mfr. No. Formulation Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Formulation Quantity Price Quantity & Availability  
MS661P1
View Documents
Thermo Scientific™
MS661P1
Purified Ab with BSA and Azide 500μL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
MS661P1ABX
View Documents
Thermo Scientific™
MS661P1ABX
Ab without BSA/Azide (Frozen) 100μL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
MS661R1
View Documents
Thermo Scientific™
MS661R1
Ready-to-use for IHC (UVLP) 1mL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
MS661P0
View Documents
Thermo Scientific™
MS661P0
Purified Ab with BSA and Azide 100μL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description

Description

Specifically detect ezrin in human, monkey, cow, kangaroo, rat, mouse and hamster samples with Thermo Scientific™ Ezrin/p81/80K/Cytovillin Ab-1, Mouse Monoclonal Antibody.

Ezrin (also known as p81, 80K, or cytovillin) is a cytoplasmic protein enriched in microvilli and other cell surface structures. Its NH2-terminal domain is associated with the plasma membrane and the COOH-terminal domain with the cytoskeleton. Ezrin has an actin-binding capability. Ezrin is expressed in epithelial cells but not in mesenchymal cells. Ezrin is also expressed by certain epithelial tumors, such as renal cell adenocarcinomas. Ezrin serves as a tyrosine kinase substrate, and is phosphorylated in EGF-stimulated cells.

Host Species: Mouse

Clone: 3C12

Isotype: IgG1

Species Reactivity: Human, Monkey, Cow, Kangaroo, Rat, Mouse and Hamster. Others not known.

Epitope: aa 362-585

Immunogen: A recombinant fragment of human ezrin corresponding to aa 362-585

Molecular Weight: 81 kDa

Positive Control: Raji cells, Lung

Cellular Localization: Membrane

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunoprecipitation (Native and denatured)

Note:

USA: RUO; Int'l: RUO

Specifications

Specifications

Ezrin/p81/80K/Cytovillin Ab-1
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Murine
Cytoskeletal Protein
A recombinant fragment of human ezrin corresponding to aa 362-585
3C12
Monoclonal
RUO
Primary
IgG1
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit