Learn More
IgG (H+L) F(ab ft.)2-Goat anti-Mouse, PE-Cyanine5, Invitrogen™ Non-distribution product as customer accommodation.
F(ab ft.)2-Goat Polyclonal Secondary Antibody
Manufacturer:  Invitrogen M35018
Catalog No. 50-113-7450
Add to cart
Description
Description
Anti-Mouse secondary antibodies are affinity-purified antibodies with well-characterized specificity for mouse immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
Specifications
Specifications
Mouse IgG (H+L)
Polyclonal
PBS with 4mg/mL BSA, sucrose and 0.1% sodium azide
F(ab′)2-Goat
IgG
500 μL
4° C, store in dark
Secondary
Mouse
Flow Cytometry
PE-Cyanine5
Liquid
Gamma Immunoglobin
Purified
RUO
Secondary Antibody
Polyclonal