Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

F-Actin Mouse anti-Human, Mouse, Chicken, Rabbit, Clone: NH3, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP261610

Catalog No. NBP261610


Add to cart

Description

Description

Actin Gamma 1 Monoclonal antibody specifically detects Actin Gamma 1 in Human, Mouse, Chicken, Rabbit samples. It is validated for Flow Cytometry, ELISA.
Specifications

Specifications

F-Actin
NH3
Unconjugated
PBS with 0.02% Sodium Azide
ACTG1
Human monocytes and the human monocyte-like cell line, U937.
0.1 ml
Primary
71
ELISA, Flow Cytometry
1 mg/mL
Flow Cytometry 1:10-1:1000, ELISA 1:100-1:2000
actin, cytoplasmic 2, DFNA26, ACT, ACTG, actin, gamma 1, BRWS2, DFNA20
Mouse
IgM Kappa
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Monoclonal
Chicken, Human, Mouse, Rabbit
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit