Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Actin Gamma 1 Antibody (NH3), Novus Biologicals™

Mouse Monoclonal Antibody has been used in 6 publications
Supplier: Novus Biologicals NB10064792SS
Description
Actin Gamma 1 Monoclonal specifically detects Actin Gamma 1 in Human, Mouse, Rat, Rabbit samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
F-Actin | |
Monoclonal | |
1.0 mg/mL | |
Western Blot 1:100-1:500, Flow Cytometry 1:10, ELISA 1:10, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500 | |
P63261 | |
ACTG1 | |
Human monocytes and U937 cell line | |
0.05 mg | |
Primary | |
Recognizes human Filamentous actin (F-actin). The binds to the N-terminal region of actin, but not to the extreme N-terminal 40 amino acids. In tissue sections the stains the cytoplasm of macrophages strongly, and gives granular, localized nuclear staining of all cell types. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgM |
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen) | |
NH3 | |
Unconjugated | |
Phosphate buffered saline with 0.09% Sodium Azide | |
actin, cytoplasmic 2, DFNA26, ACT, ACTG, actin, gamma 1, BRWS2, DFNA20 | |
Mouse | |
IgM purified | |
RUO | |
71 | |
Human, Mouse, Rat, Rabbit | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction