Learn More
CERILLIANT™ Fenproporex Hydrochloride Solution, 1 mg/mL in Methanol (as free base), Sold by MilliporeSigma™ Supelco™
Certified reference material
Manufacturer: MilliporeSigma Supelco F0561ML
Description
Fenproporex, also known as Perphoxene and used as an appetite suppressant for the treatment of obesity, is a stimulant drug of the phenethylamine and amphetamine classes that produces amphetamine as a metabolite. Fenproporex is designated a Schedule IV controlled substance in the US pursuant to the Controlled Substances Act and is also listed as banned substance by the World Anti-Doping Agency. This Snap-N-Spike™ Reference Solution is suitable for use as starting material in calibrators or controls for a variety of LC/MS or GC/MS applications from forensic analysis and urine drug testing and to clinical toxicology and sports testing.Specifications
67-56-1 | |
9.7°C (49.5°F) - closed cup | |
C12H16N2 · HCl | |
1 mL |
1 mg/mL in methanol (as free base) | |
224.73 | |
Ampule | |
-20°C |
Safety and Handling
H331-H370
H311
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
CERILLIANT is a registered trademark of Cerilliant Corporation