Learn More
CERILLIANT™ Fluticasone Impurity Mixture Solution, 20 μg/mL (0.05% H3PO4 in Acetonitrile, Sold by MilliporeSigma™ Supelco™
Certified reference material
Manufacturer:  MilliporeSigma Supelco F0611ML
Catalog No. 01-001-383
Due to product restrictions, please to purchase or view availability for this product.
Description
Description
Fluticasone belongs to the group of drugs called inhaled corticosteroids (ICSs) which are used in the management of chronic obstructive pulmonary disease (COPD). It does so by increasing the post bronchodilator forced expiratory volume in 1s (FEV1), thereby decreasing the number of acute episodes of worsening symptoms. Along with long-acting β2-agonists (LABA) such as salmeterol, it is used to reduce the frequency of exacerbations among asthma patients.
Specifications
Specifications
75-05-8,80474-14-2
2°C (35.6°F) - closed cup
C25H29F3O5S
1 mL
20 μg/mL (0.05% H3PO4 in acetonitrile
498.55
Ampule
-20°C
Safety and Handling
Safety and Handling
H332-H319
H312
SDS
Documents
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
CERILLIANT is a registered trademark of Cerilliant Corporation