Learn More
Thermo Scientific™ F1-ClipTip™ Fixed Volume Single Channel Pipettes, PROMO
With the added confidence of ClipTip technology, these pipettes are ideal for use in set protocols requiring a specific volume while providing low attachment and ejection forces.
Manufacturer:  Thermo Scientific™ 4651270NPM
 View more versions of this product
Catalog No. 14-488-030NPM
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description
Description
  • Worry-free pipetting is finally here with the only tip that clips
  • Sealed in security of ClipTip Technology
  • Eliminate the frustration of banging tips on your pipette
  • With a light touch, each tip is clipped securely on the pipette
  • Reduce the risk of loose or leaking tips
  • When ClipTip pipette tips are on, they are sealed and will not loosen. The ClipTip pipetting system remains sealed until you decide to eject tips, even in the most comprehensive applications, including multiple tip touches and mixing cycles
  • Enable consistent and reproducible pipetting from user to user
  • Higher-quality results and more efficient research
  • Ergonomically In-touch
  • Lightweight handle design combined with low plunger, tip attachment and ejection forces help reduce the risk of repetitive strain injuries
  • Secure Volume Lock
  • The secure volume setting prevents undesirable volume drift during operation. Simply lift the plunger up to set volume and press down to lock in place with minimum effort
  • Adjustable Comfort
  • The 120° adjustable finger rest – for both right- and left-handed operator comfort – allows the hand to relax during pipetting cycles
  • Improved small volume pipetting
  • Life-time Warranty
Specifications
Specifications
1000 μL
ClipTip 1000
F1-ClipTip Fixed Volume