Thermo Scientific™ F1-ClipTip™ GLP Pipette Kits
Transform daily pipetting. Innovative Thermo Scientific™ ClipTip™ interlocking technology ensures a complete seal with minimal tip attachment and ejection force.
Manufacturer: Thermo Scientific™ 4701140
 View more versions of this product
Catalog No. 14-488-039
 N/A
N/A
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.  View substitute products »
Disclaimers:
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
    Specifications
    Model F1-ClipTip™
    Volume (Metric) Range 1 to 1,000μL
    Color Code Multiple,Multiple
    Certifications/Compliance The F1-ClipTip pipetting system is CE-marked in accordance with IVD Directive in Europe. Manufactured according to ISO 9001, ISO 14001 and ISO 13485.
    ClipTip F1
    Description GLP Kit 1
    Volume Range (Metric) 1 to 1000µL
    Transform daily pipetting with Thermo Scientific™ F1-ClipTip™ GLP Pipette Kits feature unique technology that ensures a complete seal with minimal tip attachment and ejection force. Achieve newfound confidence knowing that once attached, your tips are locked firmly in place, and will not loosen or fall off regardless of application pressure. The innovative three interlocking clip design ensures the tip is held securely on the F1-ClipTip pipette until—and only until—it is released.
    Features:
    • Tip is locked and sealed in place until ejected
    • Complete seal to secure the sample volume in each tip
    • Tips are attached and ejected with a light touch
    Cutting-Edge Design:
    • Offering low attachment, ejection and plunger forces for safe and comfortable use
    • Lightweight design protects user from the risk of Repetitive Strain Injuries (RSI)
    • 120° adjustable finger rest—for both right- and left-handed operator comfort—allows the hand to relax during pipetting cycles
    Effective Functionality:
    • 150% increase in air boost to ensure efficient delivery of micro-volumes and prevent capillary action†
    • Innovative gearing mechanism produces more downward force with less effort, releasing tips with a gentle push
    • Secure volume setting prevents undesirable volume drift during operation
    Kit 1 includes:
    • F1-ClipTip 1-10µL single channel
    • F1-ClipTip 10-100µL single channel
    • F1-ClipTip 100-1000µL single channel
    • F-Series Stand
    • ClipTip 20 rack
    • ClipTip 200 rack
    • ClipTip 1000 rack
    • 25mL reservoir
    • 25mL divided reservoir
    • 100mL divided reservoir
    Kit 2 includes:
    • F1-ClipTip 2-20µL single channel
    • F1-ClipTip 20-200µL single channel
    • F1-ClipTip 100-1000µL single channel
    • F1-ClipTip 30-30µL 8-channel
    • F-Series Stand
    • ClipTip 20 rack
    • ClipTip 200 rack
    • ClipTip 300 rack
    • ClipTip 1000 rack
    • 25mL reservoir
    • 25mL divided reservoir
    • 100mL divided reservoir
    Ordering Information:
    † Available in 50µL models and below
  • Description & Specifications
  • Promotions
Related Products
Product Content Feedback