Thermo Scientific™ F1-ClipTip™ Multichannel Pipettes

Ensure a complete seal on every channel with minimal tip attachment and ejection force using these multichannel pipetters, which feature an innovative locking technology.

Manufacturer: Thermo Scientific™ 4661170

 View more versions of this product

Catalog No. 14-488-037

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.  View substitute products »


    Specifications

    Compatible Tips F1-ClipTip 200
    Model F1-ClipTip
    For Use With (Equipment) F-1 ClipTip Model 200
    Number of Channels 12-channel
    Accuracy 1.3 to 5%
    Certifications/Compliance The F1-ClipTip pipetting system is CE-marked in accordance with IVD Directive in Europe. Manufactured according to ISO 9001, ISO 14001 and ISO 13485.
    Increments 0.2µL
    Precision 0.5 to 2%
    Color Code Yellow,Yellow
    Volume (Metric) Range 10 to 100µL
    Volume Range (Metric) 10 to 100µL
    Description 12-channel
    ClipTip 200

    Ensure a complete seal on every channel with minimal tip attachment and ejection force using Thermo Scientific™ F1-ClipTip™ Pipettes, which feature innovative ClipTip interlocking technology. Once attached, tips are locked firmly in place and will not loosen or fall off regardless of application pressure. The innovative three interlocking clip design ensures the tip is held securely on the F1-ClipTip pipette until it is released.

    Features

    • Tip is locked and sealed in place until ejected—no dropped tips that waste valuable samples
    • Complete seal to secure the sample volume in each tip enhanced accuracy and precision
    • Tips are attached and ejected with a light touch—eliminates the frustration of banging tips on the pipette

    Ergonomic Design

    • Offering low attachment, ejection and plunger forces for safe and comfortable use

    Remarkably Lightweight

    • Modern design protects user from the risk of Repetitive Strain Injuries (RSI)

    Adjustable Comfort

    • The 120° adjustable finger rest—for both right- and left-handed operator comfort—allows the hand to relax during pipetting cycles

    Super Blow-out

    • 150% increase in air boost to ensure efficient delivery of micro-volumes and prevent capillary action†

    Soft-touch Tip Ejection

    • Innovative gearing mechanism produces more downward force with less effort, releasing tips with a gentle pus

    Volume Lock Adjustment

    • The secure volume setting prevents undesirable volume drift during operation

    Industry-leading Lifetime Warranty

    • With convenient online registration

    † Available in 50µL models and below

  • Description & Specifications
  • Accessories

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit