Learn More
Thermo Scientific F1-ClipTip Multichannel Pipettes
Rely on ClipTip technology to lock tips firmly in place and ensure a complete seal on every channel with minimal tip attachment and ejection forces. Available in 8- or 12-channel models.
Manufacturer: Thermo Scientific 4661110N
Description
ClipTip technology locks tips firmly in place and ensures a complete seal on every channel with minimal tip attachment and ejection forces using Thermo Scientific F1-ClipTip Multichannel Pipettes. Newly enhanced volume adjustment allows for a silent, effortless and ergonomic user experience.
Specifications
ClipTip 20 | |
0.02μL | |
8 |
F1-ClipTip | |
Pink | |
1 to 10 μL |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.