Learn More
Thermo Scientific™ F1-ClipTip™ Multichannel Pipettes Available on GSA/VA Contract for Federal Government customers only.
Rely on ClipTip technology to lock tips firmly in place and ensure a complete seal on every channel with minimal tip attachment and ejection forces. Available in 8- or 12-channel models.
Manufacturer:  Thermo Scientific™ 4661150N
 View more versions of this product
Catalog No. 14-488-035N
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description
Description
ClipTip technology locks tips firmly in place and ensures a complete seal on every channel with minimal tip attachment and ejection forces using Thermo Scientific™ F1-ClipTip™ Multichannel Pipettes. Newly enhanced volume adjustment allows for a silent, effortless and ergonomic user experience.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
ClipTip 20
0.02μL
12
F1-ClipTip
Pink
1 to 10 μL