Learn More
Thermo Scientific™ F1-ClipTip™ Multichannel Pipettes: PROMO Available on GSA/VA Contract for Federal Government customers only.
Ensure a complete seal on every channel with minimal tip attachment and ejection force using these multichannel pipetters, which feature an innovative locking technology.
Manufacturer:  Thermo Scientific™ P4661150
 View more versions of this product
Catalog No. 14-488-035TI
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description
Description
Features
  • Tip is locked and sealed in place until ejected—no dropped tips that waste valuable samples
  • Complete seal to secure the sample volume in each tip enhanced accuracy and precision
  • Tips are attached and ejected with a light touch—eliminates the frustration of banging tips on the pipette
  • Ergonomic Design
  • Offering low attachment, ejection and plunger forces for safe and comfortable use
  • Remarkably Lightweight
  • Modern design protects user from the risk of Repetitive Strain Injuries (RSI)
  • Adjustable Comfort
  • The 120° adjustable finger rest—for both right- and left-handed operator comfort—allows the hand to relax during pipetting cycles
  • Super Blow-out
  • 150% increase in air boost to ensure efficient delivery of micro-volumes and prevent capillary action†
  • Soft-touch Tip Ejection
  • Innovative gearing mechanism produces more downward force with less effort, releasing tips with a gentle pus
  • Volume Lock Adjustment
  • The secure volume setting prevents undesirable volume drift during operation
  • Industry-leading Lifetime Warranty
  • With convenient online registration
  • † Available in 50µL models and below
    This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
    Specifications
    Specifications
    2.4 to 12%
    20
    12-channel
    1.6 to 8%
    F1-ClipTip™
    1 to 10 μL
    The F1-ClipTip pipetting system is CE-marked in accordance with IVD Directive in Europe. Manufactured according to ISO 9001, ISO 14001 and ISO 13485.
    F1-ClipTip 20
    0.02μL
    Pink
    12
    Documents
    Promotions
    Promotions
    • Thermo Scientific
      Upgrade any pipette to a Thermo Scientific F1-ClipTip Manual Pipette and receive $129 off the price of a single channel pipette or $212 off the price of a multichannel one.
      12-31-2019