Learn More
Thermo Scientific F1-ClipTip Multichannel Pipettes, PROMO
Rely on ClipTip technology to lock tips firmly in place and ensure a complete seal on every channel with minimal tip attachment and ejection forces.
Manufacturer: Thermo Scientific 4661130NPM
Description
- Worry-free pipetting is finally here with the only tip that clips
- Sealed in security of ClipTip Technology
- Eliminate the frustration of banging tips on your pipette
- With a light touch, each tip is clipped securely on the pipette
- Reduce the risk of loose or leaking tips
- When ClipTip pipette tips are on, they are sealed and will not loosen. The ClipTip pipetting system remains sealed until you decide to eject tips, even in the most comprehensive applications, including multiple tip touches and mixing cycles
- Enable consistent and reproducible pipetting from user to user
- Higher-quality results and more efficient research
- Ergonomically In-touch
- Lightweight handle design combined with low plunger, tip attachment and ejection forces help reduce the risk of repetitive strain injuries
- Secure Volume Lock
- The secure volume setting prevents undesirable volume drift during operation
- Simply lift the plunger up to set volume and press down to lock in place with minimum effort
- Adjustable Comfort
- The 120° adjustable finger rest for both right- and left-handed operator comfort allows the hand to relax during pipetting cycles
- Improved small volume pipetting
- Life-time Warranty
- With convenient online registration, and may vary by country
Specifications
ClipTip 200 | |
F1-ClipTip | |
0.2 μL | |
8 |