Learn More
Thermo Scientific™ F1-ClipTip™ Variable Volume Single Channel Pipettes Available on GSA/VA Contract for Federal Government customers only.
ClipTip technology locks tips firmly to the pipette with a complete seal so they will not loosen or fall off. Lightweight handle design combined with minimal tip attachment and ejection forces.
Manufacturer:  Thermo Scientific™ 4641180N
 View more versions of this product
Catalog No. 14-488-014N
Add to cart
Description
Description
ClipTip technology locks tips firmly to the pipette with a complete seal so they will not loosen or fall off. Lightweight handle design combined with minimal tip attachment and ejection forces will increase the conform in pipetting. Feel the difference.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
ClipTip 20
0.02μL
2 to 20 μL
F1-ClipTip
Pink