Learn More
Thermo Scientific™ F1-ClipTip™ Variable Volume Single Channel Pipettes: PROMO Available on GSA/VA Contract for Federal Government customers only.
Streamline daily pipetting with innovative Thermo Scientific™ ClipTip™ interlocking technology, which ensures a complete seal with minimal tip attachment.
Manufacturer:  Thermo Scientific™ P4641190
 View more versions of this product
Catalog No. 14-488-015TI
Add to cart
Description
Description
Innovative ClipTip interlocking tip interface technology
  • Tip is locked and sealed in place until ejected— no dropped tips that waste valuable samples
  • Complete seal to secure the sample volume in each tip for enhanced accuracy and precision
  • Tips are attached and ejected with a light touch— eliminates the frustration of banging tips on the pipette
  • Ergonomic design
  • Offering low attachment, ejection and plunger forces for safe and comfortable use
  • Remarkably lightweight
  • Modern design protects user from the risk of Repetitive Strain Injuries (RSI)
  • Adjustable comfort
  • The 120° adjustable finger rest—for both right- and left-handed operator comfort—allows the hand to relax during pipetting cycles
  • Super blow-out
  • 150% increase in air boost to ensure efficient delivery of micro-volumes and prevent capillary action†
  • Soft-touch tip ejection
  • Innovative gearing mechanism produces more downward force with less effort, releasing tips with a gentle push
  • Volume Lock Adjustment
  • The secure volume setting prevents undesirable volume drift during operation
  • Industry-leading lifetime warranty
  • With convenient online registration
  • Ordering Information:
    † Available in 50µL models and below
    Compatible with:
    Thermo Scientific ClipTip pipette tips.
    This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
    Specifications
    Specifications
    0.6 to 3%
    50
    0.1μL
    Violet
    5 to 50μL
    The F1-ClipTip pipetting system is CE-marked in accordance with IVD Directive in Europe. Manufactured according to ISO 9001, ISO 14001 and ISO 13485.
    F1-ClipTip 50
    0.3 to 2.5%
    F1-ClipTip™
    Documents
    Promotions
    Promotions
    • Thermo Scientific
      Upgrade any pipette to a Thermo Scientific™ F1-ClipTip Manual Pipette and receive $129 off the price of a single channel pipette or $212 off the price of a multichannel one.
      12-31-2019
    Provide Content Correction
    We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
    Product Title
    Cancel Submit
    Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.