Learn More
F4/80 Rat anti-Mouse, Clone: CI:A3-1, Novus Biologicals™
Rat Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP2616130.1MG
 View more versions of this product
Catalog No. NBP2616131
Add to cart
Description
Description
F4/80 Monoclonal antibody specifically detects F4/80 in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry.
Specifications
Specifications
F4/80
CI:A3-1
Flow Cytometry 1:10-1:1000, Immunohistochemistry 1:10-1:500
Purified
Adgre1
Thioglycollate stimulated peritoneal macrophages of mouse origin.
Protein A purified
Immune System Diseases, Immunology, Innate Immunity, Myeloid derived Suppressor Cell
Primary
13733
Mouse
Flow Cytometry, Immunohistochemistry
Unconjugated
PBS with 0.02% Proclin 300. with No Preservative
cell surface glycoprotein F4/80, DD7A5-7, EGF-like module containing, mucin-like, hormone receptor-like sequence 1, EGF-like module receptor 1, EGF-like module-containing mucin-like hormone receptor-like 1, EGF-TM7, EMR1 hormone receptor, F4/80, Gpf480, Ly71, lymphocyte antigen 71, Mouse EMR1, Rat EMR1, TM7LN3
Rat
IgG1 Kappa
0.1 mg
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Monoclonal
This antibody recognises the mouse F4/80 antigen, a 160kD glycoprotein, expressed by murine macrophages.