Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
F4/80 Rat anti-Mouse, DyLight 755, Clone: CI:A3-1, Novus Biologicals™
Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP261613IR
Description
F4/80 Monoclonal specifically detects F4/80 in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry.Specifications
F4/80 | |
Monoclonal | |
DyLight 755 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
Adgre1 | |
Thioglycollate stimulated peritoneal macrophages of mouse origin. | |
0.1 mL | |
Primary | |
This antibody recognises the mouse F4/80 antigen, a 160kD glycoprotein, expressed by murine macrophages. | |
Store at 4°C in the dark. | |
IgG1 κ |
Flow Cytometry, Immunohistochemistry | |
CI:A3-1 | |
Flow Cytometry, Immunohistochemistry | |
ADGRE1, Adhesion G protein-coupled receptor E1, cell surface glycoprotein F4/80, DD7A5-7, EGF-like module containing, mucin-like, hormone receptor-like sequence 1, EGF-like module receptor 1, EGF-like module-containing mucin-like hormone receptor-like 1, EGF-TM7, EMR1 hormone receptor, F4/80, Gpf480, Ly71, lymphocyte antigen 71, Mouse EMR1, Rat EMR1, TM7LN3 | |
Rat | |
Protein A purified | |
Immune System Diseases, Immunology, Innate Immunity, Myeloid derived Suppressor Cell | |
2015 | |
Mouse | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction