Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

F4/80 Rat anti-Mouse, Janelia Fluor™ 646, Clone: CI:A3-1, Novus Biologicals™

Rat Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP261613JF646

Catalog No. N261613J646


Add to cart

Description

Description

F4/80 Monoclonal antibody specifically detects F4/80 in Mouse samples. It is validated for Flow Cytometry.
Specifications

Specifications

F4/80
CI:A3-1
Flow Cytometry
Conjugated, Purified
Adgre1
Thioglycollate stimulated peritoneal macrophages of mouse origin.
Protein A purified
Immune System Diseases, Immunology, Innate Immunity, Myeloid derived Suppressor Cell
Primary
13733.0
Mouse
Flow Cytometry
Janelia Fluor 646
50mM Sodium Borate with 0.05% Sodium Azide
ADGRE1, cell surface glycoprotein F4/80, DD7A5-7, EGF-like module containing, mucin-like, hormone receptor-like sequence 1, EGF-like module receptor 1, EGF-like module-containing mucin-like hormone receptor-like 1, EGF-TM7, EMR1 hormone receptor, F4/80, Gpf480, Ly71, lymphocyte antigen 71, Mouse EMR1, Rat EMR1, TM7LN3
Rat
IgG1 Kappa
0.1 mL
Store at 4°C in the dark.
Monoclonal
This antibody recognises the mouse F4/80 antigen, a 160kD glycoprotein, expressed by murine macrophages.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit