Learn More
F4/80 Rat anti-Mouse, PE-CF594, Clone: T45-2342, BD Horizon™
Rat Monoclonal Antibody
Manufacturer:  BD Biosciences 565613
Catalog No. BDB565613
Add to cart
Description
Description
F4/80 Monoclonal antibody specifically detects F4/80 in Mouse samples. It is validated for Flow cytometry.
Specifications
Specifications
F4/80
T45-2342
PE-CF594
Purified
Rat
Rat WI
RUO
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon™ PE-CF594 under optimum conditions
Monoclonal
Flow Cytometry
0.2mg/mL
Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Gpf480; F480; Emr1; Ly71; DD7A5-7; EGF-TM7; TM7LN3
Mouse F4/80 Recombinant Protein
50μg
Cell Biology
Primary
Murine
Documents
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
For Reseach Use Only.