Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

F4/80 Rat anti-Mouse, PE, Clone: BM8, Invitrogen™ Non-distribution product as customer accommodation.

Rat Monoclonal Antibody

Manufacturer:  Invitrogen MF480043

Catalog No. 50-113-8113


Add to cart

Description

Description

F4/80 antigen is a 160 kDa cell surface glycoprotein that is a member of the EGF TM7 family of proteins and shares 68% overall amino acid identity with human EGF module containing mucin like hormone receptor 1 (EMR1). Expression of F4/80 is heterogeneous and is reported to vary during macrophage maturation and activation. The F4/80 antigen is expressed on a wide range of mature tissue macrophages including Kupffer cells, Langerhans, microglia, macrophages located in the gut lamina propria, peritoneal cavity, lung, thymus, bone marrow stroma and macrophages in the red pulp of the spleen. F4/80 expression has also been reported on a subpopulation of dendritic cells but is absent from macrophages located in T cell areas of the spleen and lymph node. The ligands and biological functions of the F4/80 antigen have not yet been determined but recent studies suggest a role for F4/80 in the generation of efferent CD8+ve regulatory T cells.
Specifications

Specifications

F4/80
Monoclonal
0.2 mg/mL
PBS with 10% sucrose, 4mg/mL BSA and 0.1% sodium azide
Q61549
EMR1, EMR1 hormone receptor, F4/80, EGF-like module receptor 1, TM7LN3
Rat
IgG2a
3 mL
4° C
Primary
13733
Flow Cytometry
BM8
PE
Adgre1
Liquid
Adgre1
mouse F4/80 which is a macrophage-restricted cell surface glycoprotein.
Purified
RUO
Antibody
Monoclonal
Mouse
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit