Learn More
F5 Rabbit anti-Human, Mouse, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 209631AP
Catalog No. 50-173-3177
Add to cart
Description
Description
Coagulation factor V (or Proaccelerin) is an essential factor of the blood coagulation cascade. This factor circulates in plasma, and is converted to the active form by the release of the activation peptide by thrombin during coagulation. This generates a heavy chain and a light chain which are held together by calcium ions. The active factor V is a cofactor that participates with activated coagulation factor X to activate prothrombin to thrombin. Defects in the Factor V gene result in either an autosomal recessive hemorrhagic diathesis or an autosomal domit form of thrombophilia, which is known as activated protein C resistance.
Specifications
Specifications
F5
0.37 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
O88783, P12259
Activated protein C cofactor, Coagulation factor V, F5, FVL, PCCF, Proaccelerin, labile factor
Rabbit
IgG
150 μL
-20°C
Primary
14067, 2153
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
F5
Liquid
F5
Peptide
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse