Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ F7 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5109703
Description
Immunogen sequence: RNLIAVLGEH DLSEHDGDEQ SRRVAQVIIP STYVPGTTNH DIALLRLHQP VVLTDHVVPL CLPERTFSER TLAFVRFSLV SGWGQLLDRG ATALELMVLN VPRLMTQDCL QQSRKVGDSP NITEYMFCAG YSDGSKDSCK GDSGGPHATH YRGTWYLTGI V.
Coagulation factor VII is a vitamin K-dependent factor essential for hemostasis. This factor circulates in the blood in a zymogen form, and is converted to an active form by either factor IXa, factor Xa, factor XIIa, or thrombin by minor proteolysis. Upon activation of the factor VII, a heavy chain containing a catalytic domain and a light chain containing 2 EGF-like domains are generated, and two chains are held together by a disulfide bond. In the presence of factor III and calcium ions, the activated factor then further activates the coagulation cascade by converting factor IX to factor IXa and/or factor X to factor Xa. Defects in F7 production can cause coagulopathy.
Specifications
F7 | |
Polyclonal | |
Unconjugated | |
F7 | |
AI132620; Cf7; Coagulation factor VII; coagulation factor VII (serum prothrombin conversion accelerator); eptacog alfa; F7; Factor 7; Factor VII heavy chain; Factor VII light chain; Factor7; FVII; FVII coagulation protein; proconvertin; Serum prothrombin conversion accelerator; SPCA | |
Rabbit | |
Affinity Chromatography | |
RUO | |
14068, 2155, 260320 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
2.86 mg/mL | |
PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
P08709, P70375, Q8K3U6 | |
F7 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 240-400 of human F7 (NP_062562.1). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction