Learn More
F8 Rabbit anti-Human, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 214851AP
Catalog No. 50-173-3445
Add to cart
Description
Description
This antibody can recognize the light chain.
This gene encodes coagulation factor VIII, which participates in the intrinsic pathway of blood coagulation; factor VIII is a cofactor for factor IXa which, in the presence of Ca+2 and phospholipids, converts factor X to the activated form Xa. This gene produces two alternatively spliced transcripts. Transcript variant 1 encodes a large glycoprotein, isoform a, which circulates in plasma and associates with von Willebrand factor in a noncovalent complex. This protein undergoes multiple cleavage events. Transcript variant 2 encodes a putative small protein, isoform b, which consists primarily of the phospholipid binding domain of factor VIIIc. This binding domain is essential for coagulant activity. Defects in this gene results in hemophilia A, a common recessive X-linked coagulation disorder.
Specifications
Specifications
F8
0.13 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P00451
AHF, Antihemophilic factor, Coagulation factor VIII, DXS1253E, F8, F8B, F8C, FVIII, HEMA, Procoagulant component
Rabbit
IgG
150 μL
-20°C
Primary
2157
Immunocytochemistry, Immunofluorescence
Unconjugated
F8
Liquid
F8
F8 Fusion Protein Ag15772
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human