Learn More
F9 Mouse anti-Human, Clone: 13F42-F6, Abnova™
Mouse monoclonal antibody raised against human F9.
Manufacturer:  Abnova Corporation MAB12651
Catalog No. 89-934-115
Add to cart
Description
Description
This gene encodes vitamin K-dependent coagulation factor IX that circulates in the blood as an inactive zymogen. This factor is converted to an active form by factor XIa, which excises the activation peptide and thus generates a heavy chain and a light chain held together by one or more disulfide bonds. The role of this activated factor IX in the blood coagulation cascade is to activate factor X to its active form through interactions with Ca+2 ions, membrane phospholipids, and factor VIII. Alterations of this gene, including point mutations, insertions and deletions, cause factor IX deficiency, which is a recessive X-linked disorder, also called hemophilia B or Christmas disease. [provided by RefSeq]
Specifications
Specifications
F9
13F42-F6
Mouse monoclonal antibody raised against human F9.
In 0.05M Na
FIX/HEMB/MGC129641/MGC129642/PTC
Mouse
IgG1, kappa
250 ug
Store at -80°C.
Aliquot to avoid repeated freezing and thawing.
Monoclonal
Human
ELISA, Immunohistochemistry (PFA fixed), Western Blot
Unconjugated
ELISA Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (10 ug/mL) Western Blot The optimal working dilution should be determined by the end user.
Liquid
F9
Native purified human F9.
IgG purification
RUO
Primary
2158