Learn More
F9 Rabbit anti-Human, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 214811AP
Catalog No. 50-173-3441
Add to cart
Description
Description
Coagulation factor IX is a single chain polypeptide with a molecular mass of 57 kDa. Normal concentrations of factor IX in human plasma is approximately 3 µg/mL. Factor IX deficiency and lack of coagulation activity is seen in patients suffering from the innate X-chromosome linked disorder, haemophilia B.
Specifications
Specifications
F9
0.34 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P00740
Christmas factor, coagulation factor IX, F9, FIX, HEMB, PTC
Rabbit
IgG
150 μL
-20°C
Primary
2158
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
F9
Liquid
F9
F9 Fusion Protein Ag15723
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human