Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eppendorf™ Rotor FA - 6 x 250 mL with QuickLock™ Rotor Lid

Ideal for the pelleting of microorganisms and mammalian cells

Manufacturer:  Eppendorf™ 5895175007

Catalog No. 05-413-901


Add to cart

Description

Description

  • Fixed-angle rotor for up to 6 x 250 mL bottles
  • 12 different adapters for broad range of additional applications
  • Max. speed 15,054 x g (10,100 rpm)
  • Eppendorf™ QuickLock™ rotor lid for fast lid locking
  • Aerosol-tight for safe centrifugation of hazardous samples
  • Low weight of only 5,450 g for stress-free work
  • Anodized aluminum for high chemical resistance
  • Rotor, lid and adapters are autoclavable to reduce risk of contamination
  • See tab 'Additional information' for overview of adapters available
  • To run this rotor, a software update is required for earlier centrifuges. Please contact your local Eppendorf organization or dealer for further information
  • Aerosol-tightness tested by Public Health England, Porton Down, UK
  • Maximum 80% of maximum filling volume
Specifications

Specifications

Rotor FA - 6 x 250
15,054 × g
Centrifuge 5910 R and 5920 R
6 × 250 mL
10,100 rpm
6
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit