Learn More
Epredia™ Fa-Tech™ Series 315 Laser Embedding Cassettes
Manufactured specifically for laser printing, the Fa-Tech™ Series 315 Embedding Cassettes have round holes to maximize fluid exchange.
$710.00
Specifications
Closure Included | No |
---|---|
Quantity | 40 Cassettes/Taped Stack, 45 Stacks/Cs. |
Pore Style | Round Holes |
Catalog Number | Mfr. No. | Color | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Color | Price | Quantity & Availability | |||||
3150118LM
|
Epredia™
3150118LM |
White |
Case of 1800 for $710.00
|
|
3150218LM
|
Epredia™
3150218LM |
Yellow |
Case of 1800 for $710.00
|
|
3150318LM
|
Epredia™
3150318LM |
Blue |
Case of 1800 for $710.00
|
|
3150418LM
|
Epredia™
3150418LM |
Pink |
Case of 1800 for $710.00
|
|
3150518LM
|
Epredia™
3150518LM |
Green |
Case of 1800 for $710.00
|
|
3150618LM
|
Epredia™
3150618LM |
Orange |
Case of 1800 for $710.00
|
|
3150718LM
|
Epredia™
3150718LM |
Gray |
Case of 1800 for $710.00
|
|
3150818LM
|
Epredia™
3150818LM |
Aqua |
Case of 1800 for $710.00
|
|
3150918LM
|
Epredia™
3150918LM |
Lilac |
Case of 1800 for $710.00
|
|
3151218LM
|
Epredia™
3151218LM |
Red |
Case of 1800 for $710.00
|
|
Description
- Fa-Tech™ Laser Cassettes are designed for use on both the NOVA™ and VEGA™ Laser Printers.
- They contain a special constituent chemical compound that carbonizes when exposed to the laser beam.
- The embedding style of cassette comes in 10 different colors: white, yellow, blue, pink, green, orange, grey, aqua, lilac and red.
- Required reusable metal cassette lids (Part number FTML315) are sold separately.
- The embedding cassette style is designed to maximize fluid exchange during processing and come in stacks held together with tape for ease of loading into Fa-Tech™ cassette hoppers for printing.
- Series 315 comes in taped stacks of 40 cassettes; 25 stacks per case.
- None of the Fa-Tech™ Laser Cassette Series are compatible with thermal tape style printers.
Specifications
No | |
40 Cassettes/Taped Stack, 45 Stacks/Cs. | |
Round Holes |