Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FA2H Antibody (OTI2E3) - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272192
Description
FA2H Monoclonal specifically detects FA2H in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
FA2H | |
Monoclonal | |
LYOPH | |
Western Blot 1:1000, Immunocytochemistry/Immunofluorescence 1:50-1:100 | |
FAAHFAXDC1, FAH1, fatty acid 2-hydroxylase, Fatty acid alpha-hydroxylase, fatty acid hydroxylase domain containing 1, FLJ25287, SCS7, spastic paraplegia 35 (autosomal recessive), SPG35 | |
Mouse | |
42.6 kDa | |
100 μg | |
Primary | |
Reconstitute with PBS (pH 7.3). To use this carrier-free antibody for conjugation experiments, another round of desalting is highly recommended. | |
Store at -20°C. Avoid freeze-thaw cycles. | |
IgG1 |
Western Blot, Immunocytochemistry, Immunofluorescence | |
OTI2E3 | |
Unconjugated | |
Lyophilized from PBS (pH 7.3) with 8% Trehalose with No Preservative | |
FA2H | |
Full length human recombinant protein of human FA2H(NP_077282) produced in HEK293T cell. | |
Affinity Purified | |
Lipid and Metabolism | |
79152 | |
Human | |
Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction