Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FAM83G Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548644

Catalog No. PA548644

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

May regulate the bone morphogenetic proteins (BMP) pathway.
Specifications

Specifications

FAM83G
Polyclonal
PBS with 0.09% sodium azide
A6ND36
Protein FAM83G, Protein associated with SMAD1, FAM83G, PAWS1
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
644815
Western Blot
Unconjugated
FAM83G
Liquid
FAM83G
KLH conjugated synthetic peptide between 794-823 amino acids from the C-terminal region of human FA83G.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit