Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FAAH Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
$496.07
Specifications
Antigen | FAAH |
---|---|
Concentration | 0.27 mg/mL |
Applications | Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-1869
|
Proteintech Group Inc
179091AP150UL |
150 μL |
Each of 1 for $496.07
|
|
|||||
Description
Degrades bioactive fatty acid amides like oleamide, the endogenous levels of cannabinoid, adamide and myristic amide to their corresponding acids, thereby serving to terminate the signaling functions of these molecules. Hydrolyzes polyunsaturated substrate adamide preferentially as compared to monounsaturated substrates.Specifications
FAAH | |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
O00519, O08914, P97612 | |
14073, 2166, 29347 | |
FAAH Fusion Protein Ag12304 | |
Primary | |
-20°C | |
FAAH |
0.27 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Adamide amidohydrolase 1, FAAH, FAAH 1, FAAH1, fatty acid amide hydrolase, Fatty acid amide hydrolase 1, Oleamide hydrolase 1 | |
FAAH | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title